Learn More
Alfa Aesar™ Acide N-benzoyl-L-glutamique, 98 %
Marque: Alfa Aesar™ H66778.14
Code nomenclature Nacres: NA.26
Informations supplémentaires : CAS : 6094-36-6 Poids : 0.02500kg
Quantité. | 25g |
---|
Identifiants chimiques
6094-36-6 | |
251.238 | |
LPJXPACOXRZCCP-VIFPVBQESA-N | |
11345724 | |
C1=CC=C(C=C1)C(=O)NC(CCC(=O)O)C(=O)O |
C12H13NO5 | |
MFCD00014028 | |
bz-glu-oh, n-benzoyl-l-glutamic acid, benzoyl-l-glutamic acid, s-2-benzamidopentanedioic acid, n-benzoylglutamic acid, unii-3591d74mxp, 2s-2-phenylformamido pentanedioic acid, +-n-benzoylglutamic acid, n-4-benzoyl-l-glutamic acid, 2s-2-phenylcarbonylamino pentanedioic acid | |
acide (2S)-2-benzamidopentanédioïque |
Spécification
N-Benzoyl-L-glutamic acid | |
C12H13NO5 | |
2622989 | |
LPJXPACOXRZCCP-VIFPVBQESA-N | |
acide (2S)-2-benzamidopentanédioïque | |
11345724 | |
Solide | |
25g |
6094-36-6 | |
MFCD00014028 | |
bz-glu-oh, n-benzoyl-l-glutamic acid, benzoyl-l-glutamic acid, s-2-benzamidopentanedioic acid, n-benzoylglutamic acid, unii-3591d74mxp, 2s-2-phenylformamido pentanedioic acid, +-n-benzoylglutamic acid, n-4-benzoyl-l-glutamic acid, 2s-2-phenylcarbonylamino pentanedioic acid | |
C1=CC=C(C=C1)C(=O)NC(CCC(=O)O)C(=O)O | |
251.238 | |
251.24 | |
98% |