missing translation for 'onlineSavingsMsg'
Learn More

N-benzyl-4-bromobenzènesulfonamide, 97 %, Thermo Scientific™

Catalog Number p-7037775
Click to view available options
Quantité:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
3609-87-8
C13H12BrNO2S
326.208
MFCD00450587
KBFYMIDQZWIKJR-UHFFFAOYSA-N
n-benzyl 4-bromobenzenesulfonamide, n-benzyl-p-bromobenzenesulfonamide, n-benzyl-4-bromobenzene-1-sulfonamide, acmc-1cmxa, n-benzyl-4-bromobenzenesulphonamide, n-benzyl-4-bromo-benzenesulfonamide, 4-bromophenyl sulfonyl benzylamine, benzenesulfonamide, n-benzyl-p-bromo, n-benzyl-4-bromo-1-benzenesulfonamide, 4-bromo-n-phenylmethyl benzenesulfonamide
571651
N-benzyl-4-bromobenzènesulfonamide
C1=CC=C(C=C1)CNS(=O)(=O)C2=CC=C(C=C2)Br
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
3609-87-8
C13H12BrNO2S
326.208
MFCD00450587
KBFYMIDQZWIKJR-UHFFFAOYSA-N
n-benzyl 4-bromobenzenesulfonamide, n-benzyl-p-bromobenzenesulfonamide, n-benzyl-4-bromobenzene-1-sulfonamide, acmc-1cmxa, n-benzyl-4-bromobenzenesulphonamide, n-benzyl-4-bromo-benzenesulfonamide, 4-bromophenyl sulfonyl benzylamine, benzenesulfonamide, n-benzyl-p-bromo, n-benzyl-4-bromo-1-benzenesulfonamide, 4-bromo-n-phenylmethyl benzenesulfonamide
571651
N-benzyl-4-bromobenzènesulfonamide
C1=CC=C(C=C1)CNS(=O)(=O)C2=CC=C(C=C2)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau N-Benzyl-4-bromobenzenesulfonamide
Quantité 250 mg
Poids de la formule 326.21
Pourcentage de pureté 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'