missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Ester éthylique de la N-Boc-L-bêta-proline, 97 %
Marque: Alfa Aesar™ H57034.03
Code nomenclature Nacres: NA.22
Informations supplémentaires : Poids : 0.00100kg
Quantité. | 1g |
---|
Identifiants chimiques
1421256-03-2 | |
243.303 | |
PKDQVUYUWUHNMG-VIFPVBQESA-N | |
1515159 | |
CCOC(=O)C1CCN(C1)C(=O)OC(C)(C)C |
C12H21NO4 | |
MFCD06204615 | |
s-1-tert-butyl 3-ethyl pyrrolidine-1,3-dicarboxylate, n-boc-l-beta-proline ethyl ester, ethyl1-boc-3-pyrrolidinecarboxylate, ethyl s-1-boc-pyrrolidine-3-carboxylate, s-1-boc-pyrrolidine-3-carboxylic acid ethyl ester, 1-o-tert-butyl 3-o-ethyl 3s-pyrrolidine-1,3-dicarboxylate, 1-tert-butyl 3-ethyl 3s-pyrrolidine-1,3-dicarboxylate | |
1-O-tert-butyl 3-O-ethyl (3S)-pyrrolidine-1,3-dicarboxylate |
Spécification
N-Boc-L-beta-proline ethyl ester | |
1421256-03-2 | |
MFCD06204615 | |
PKDQVUYUWUHNMG-VIFPVBQESA-N | |
1-O-tert-butyl 3-O-ethyl (3S)-pyrrolidine-1,3-dicarboxylate | |
1515159 | |
97% |
1g | |
C12H21NO4 | |
s-1-tert-butyl 3-ethyl pyrrolidine-1,3-dicarboxylate, n-boc-l-beta-proline ethyl ester, ethyl1-boc-3-pyrrolidinecarboxylate, ethyl s-1-boc-pyrrolidine-3-carboxylate, s-1-boc-pyrrolidine-3-carboxylic acid ethyl ester, 1-o-tert-butyl 3-o-ethyl 3s-pyrrolidine-1,3-dicarboxylate, 1-tert-butyl 3-ethyl 3s-pyrrolidine-1,3-dicarboxylate | |
CCOC(=O)C1CCN(C1)C(=O)OC(C)(C)C | |
243.303 | |
243.3 |