missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAGS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21566-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
NAGS is a mitochondrial enzyme that catalyzes the formation of N-acetylglutamate (NAG) from glutamate and acetyl coenzyme-A. NAG is a cofactor of carbamyl phosphate synthetase I (CPSI), the first enzyme of the urea cycle in mammals. Deficiencies in N-acetylglutamate synthase have been associated with hyperammonemia.The N-acetylglutamate synthase gene encodes a mitochondrial enzyme that catalyzes the formation of N-acetylglutamate (NAG) from glutamate and acetyl coenzyme-A. NAG is a cofactor of carbamyl phosphate synthetase I (CPSI), the first enzyme of the urea cycle in mammals. This gene may regulate ureagenesis by altering NAG availability and, thereby, CPSI activity. Deficiencies in N-acetylglutamate synthase have been associated with hyperammonemia.Spécification
NAGS | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N159, Q8R4H7 | |
AGAS, Amino acid acetyltransferase, ARGA, N acetylglutamate synthase, NAGS | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
162417, 217214, 303563 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
NAGS | |
Liquid | |
Nags | |
NAGS Fusion Protein Ag16073 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |