missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Napsin A Mouse, Clone: NAPSA/1238, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 9476-MSM2-P1
Informations supplémentaires : Poids : 0.01000kg
Spécification
Napsin A | |
Cytoplasmic | |
NAPSA/1238 | |
Unconjugated | |
XP_016883001 | |
Mouse | |
37kDa | |
Protein A/G Purified | |
RUO | |
9476 | |
IgG1 κ |
Western Blot | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
NAPSA | |
Recombinant human Napsin-A protein fragment (aa189-299) | |
Lung adenocarcinoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |