missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFB5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23855-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
NDUFB5 is a subunit of the multisubunit NADH:ubiquinone oxidoreductase (complex I). Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. The protein encoded by this gene is a subunit of the multisubunit NADH:ubiquinone oxidoreductase (complex I). Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.Spécification
NDUFB5 | |
Polyclonal | |
Unconjugated | |
NDUFB5 | |
CI SGDH, Complex I SGDH, NDUFB5, SGDH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4711, 66046 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43674, Q9CQH3 | |
NDUFB5 | |
NDUFB5 Fusion Protein Ag20907 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |