missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK6 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10378-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The Aspergillus nidulans ′never in mitosis A′ (NIMA) gene encodes a serine/threonine kinase that controls initiation of mitosis. NIMA-related kinases (NEKs) are a group of protein kinases that are homologous to NIMA. Evidence suggests that NEKs perform functions similar to those of NIMA. NEK6 encodes a NEK type protein kinase, which contains a C-terminal NIMA-like catalytic domain.Spécification
NEK6 | |
Polyclonal | |
Unconjugated | |
NEK6 | |
NEK6, NimA related protein kinase 6, Protein kinase SID6 1512, SID6 1512 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10783, 360161, 59126 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P59895, Q9ES70, Q9HC98 | |
Nek6 | |
NEK6 Fusion Protein Ag0266 | |
20 μL | |
Primary | |
Rat, Human, Mouse, Non-human Primate | |
Antibody | |
IgG |