missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEURL3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16648-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
E3 ubiquitin-protein ligase. Seems to utilize UBE2E1. In vitro, generates polyubiquitin chains via non-canonical lysine residues suggesting that it is not involved in tagging substrates for proteasomal degradation.Spécification
NEURL3 | |
Polyclonal | |
Unconjugated | |
NEURL3 | |
LINCR, NEURL3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
214854, 93082 | |
-20°C | |
Liquid |
Immunoprecipitation | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8CJC5, Q96EH8 | |
NEURL3 | |
NEURL3 Fusion Protein Ag10035 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |