missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nidogen 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13530-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Basement membranes, which are composed of type IV collagens, laminins, perlecan, and nidogen, are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis. Basement membranes, which are composed of type IV collagens, perlecan, and nidogen, are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis.Spécification
Nidogen 2 | |
Polyclonal | |
Unconjugated | |
NID2 | |
NID 2, NID2, Nidogen 2, nidogen 2 (osteonidogen), Osteonidogen | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18074, 22795, 302248 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B5DFC9, O88322, Q14112 | |
Nid2 | |
Nidogen 2 Fusion Protein Ag4424 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |