missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NINJ2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14085-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Homophilic cell adhesion molecule that promotes axonal growth. May play a role in nerve regeneration and in the formation and function of other tissues.Spécification
NINJ2 | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9JHE8, Q9JL89, Q9NZG7 | |
Nerve injury induced protein 2, NINJ2, ninjurin 2 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
29862, 4815, 59115 |
Western Blot | |
Unconjugated | |
NINJ2 | |
Liquid | |
Ninj2 | |
NINJ2 Fusion Protein Ag5215 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |