missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NME3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | NME3 |
---|---|
Concentration | 0.21 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16834184
|
Proteintech
15136-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16824184
|
Proteintech
15136-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
NME3 mRNA is preferentially expressed at early stages of myeloid differentiation of highly purified CD34(+) cells. Its constitutive expression in a myeloid precursor line, which is growth-factor dependent for both proliferation and differentiation, results in inhibition of granulocytic differentiation induced by granulocyte colony-stimulating factor and causes apoptotic cell death. These results appear consistent with a role for the NME3 gene in normal hematopoiesis and raise the possibility that its overexpression contributes to differentiation arrest, a feature of blastic transformation in chronic myelogenous leukemia.Spécification
NME3 | |
Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q13232, Q9WV85 | |
4832, 79059, 85269 | |
NME3 Fusion Protein Ag7284 | |
Primary | |
-20°C | |
NME3 |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
c371H6.2, DR nm23, KIAA0516, NDK 3, NDP kinase 3, NDPK C, NDPKC, NME3 | |
NME3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |