missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Noggin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14772-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Noggin is involved in numerous developmental processes, such as neural tube fusion and joint formation. The morphogenesis of organs is initiated by a downgrowth from a layer of epithelial stem cells. This process is achieved through the receipt of signals from 1) a WNT protein (WNT3A) to stabilize beta-catenin; and 2) Noggin, which is a bone morphogenetic protein inhibitor. Noggin mutations in unrelated families with proximal symphalangism (SYM1) and multiple synostoses syndrome (SYNS1) have been identified, which have multiple joint fusion as their principal defect.Spécification
Noggin | |
Polyclonal | |
Unconjugated | |
NOG | |
NOG, Noggin, SYM1, SYNS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18121, 25495, 9241 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97466, Q13253, Q62809 | |
Nog | |
Noggin Fusion Protein Ag6586 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |