missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPAS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13701-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
NPAS1 is a member of the basic helix-loop-helix (bHLH)-PAS family of transcription factors. Studies of a related mouse gene suggest that it functions in neurons. The exact function of this gene is unclear, but it may play protective or modulatory roles during late embryogenesis and postnatal development.Spécification
NPAS1 | |
Polyclonal | |
Unconjugated | |
NPAS1 | |
bHLHe11, Member of PAS protein 5, MOP5, neuronal PAS domain protein 1, Neuronal PAS1, NPAS1, PASD5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18142, 308387, 4861 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P97459, Q99742 | |
Npas1 | |
NPAS1 Fusion Protein Ag4620 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |