Learn More
NPR1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 55116-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Guanylyl cyclases, catalyzing the production of cGMP from GTP, are classified as soluble and membrane forms. The membrane guanylyl cyclases, often termed guanylyl cyclases A through F, form a family of cell-surface receptors with a similar topographic structure: an extracellular ligand-binding domain, a single membrane-spanning domain, and an intracellular region that contains a protein kinase-like domain and a cyclase catalytic domain. GC-A and GC-B function as receptors for natriuretic peptides; they are also referred to as atrial natriuretic peptide receptor A and type B. Also see NPR3, which encodes a protein with only the ligand-binding transmembrane and 37-amino acid cytoplasmic domains. NPR1 is a membrane-bound guanylate cyclase that serves as the receptor for both atrial and brain natriuretic peptides and BNP, respectively).Spécification
NPR1 | |
Polyclonal | |
Unconjugated | |
NPR1 | |
ANP A, ANPa, ANPR A, ANPRA, GC A, Guanylate cyclase A, GUC2A, GUCY2A, NPR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4881 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P16066 | |
NPR1 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |