missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRBP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21549-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Nrbp2 may regulate apoptosis of neural progenitor cells during their differentiation.Spécification
NRBP2 | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q91V36, Q9NSY0 | |
NRBP2, pp9320, TRG 16, TRG16 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
223649, 340371, 680451 |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
NRBP2 | |
Liquid | |
NRBP2 | |
NRBP2 Fusion Protein Ag16215 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |