missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15630-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Cleaves peptide substrates on the N-terminus of arginine residues in dibasic pairs.Spécification
NRD1 | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43847, P47245, Q8BHG1 | |
hNRD1, hNRD2, N arginine dibasic convertase, Nardilysin, NRD C, NRD convertase, NRD1 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
230598, 25499, 4898 |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
NRDC | |
Liquid | |
Nrd1, NRDC | |
NRD1 Fusion Protein Ag7923 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |