missing translation for 'onlineSavingsMsg'
Learn More

NSE Mouse anti-Human, Mouse, Porcine, Rat, Clone: 6F8G3, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 66150-1-IG-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16827765

  • 325.38€ / 150µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Neuron specific enolase (NSE, ENO1, ENO2, ENO3) is an enzyme that catalyzes the conversion of 2-phosphoglycerate to phosphoenolpyruvate in the glycolytic pathway, and the reverse reaction in gluconeogenesis. NSE has a high stability in biological fluids and can easily diffuse to the extracellular medium and cerebrospinal fluid (CSF) when neuronal membranes are injured. NSE is one of three mammalian enolases, which are also known as ENO1, ENO2, and ENO3 or alternately as enolase alpha, beta and gamma. The alpha-subunit is expressed in most tissues, the beta-subunit only in muscle, and the gamma-subunit is expressed primarily in neurons, in normal and in neoplastic neuroendocrine cells. Co-expression of NSE and chromogranin A is common in neuroendocrine neoplasms. Since neurons require a great deal of energy, they are very rich in glycolytic enzymes such a GAPDH and NSE. Antibodies to NSE protein are useful to identify neuronal cell bodies, developing neuronal lineage and neuroendocrine cells.
Spécification

Spécification

NSE
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07323, P09104, P17183
ENO2
NSE Fusion Protein Ag19106
150 μL
Primary
Rat, Porcine, Mouse, Human
Antibody
IgG1
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
6F8G3
Unconjugated
ENO2
ENO2, Enolase 2, enolase 2 (gamma, neuronal), Gamma enolase, Neural enolase, Neuron specific enolase, NSE
Mouse
Protein G
RUO
100157750, 13807, 2026, 24334
-20°C
Liquid
Vidéos
FDS
Documents
Certificats
Promotions

Promotions