missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT5DC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24102-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
While the exact function of the protein encoded by this gene is not known, it belongs to the 5′(3′)-deoxyribonucleotidase family.Spécification
NT5DC1 | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5TFE4 | |
C6orf200, LP2642, NT5C2L1, NT5DC1 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
221294 |
Immunofluorescence, Immunocytochemistry, Western Blot | |
Unconjugated | |
NT5DC1 | |
Liquid | |
NT5DC1 | |
NT5DC1 Fusion Protein Ag20916 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |