missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15068-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
YBX1 binds to splice sites in pre-mRNA and regulates splice site selection. This protein binds and stabilizes cytoplasmic mRNA and contributes to the regulation of translation by modulating the interaction between the mRNA and eukaryotic initiation factors. It binds to promoters that contain a Y-box (5′-CTGATTGGCCAA-3′), such as HLA class II genes. It regulates the transcription of numerous genes and promotes separation of DNA strands that contain mismatches or are modified by cisplatin. It has endonucleolytic activity and can introduce nicks or breaks into double-stranded DNA (in vitro), and it may play a role in DNA repair.Spécification
NUDT9 | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5XIG0, Q8BVU5, Q9BW91 | |
ADP ribose diphosphatase, ADP ribose phosphohydrolase, ADPR PPase, Nudix motif 9, NUDT10, NUDT9, UNQ3012/PRO9771 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
305149, 53343, 74167 |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
NUDT9 | |
Liquid | |
NUDT9 | |
NUDT9 Fusion Protein Ag7075 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Human, Rat |