missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP62 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13916-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
SDS3 is a subunit of the histone deacetylase.Spécification
NUP62 | |
Polyclonal | |
Unconjugated | |
NUP62 | |
62 kDa nucleoporin, IBSN, Nuclear pore glycoprotein p62, nucleoporin 62kDa, Nucleoporin Nup62, NUP62, p62, SNDI | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18226, 23636, 65274 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P17955, P37198, Q63850 | |
Nup62 | |
NUP62 Fusion Protein Ag4912 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |