missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP93 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13077-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. This gene encodes a nucleoporin protein that localizes both to the basket of the pore and to the nuclear entry of the central gated channel of the pore. The encoded protein is a target of caspase cysteine proteases that play a central role in programmed cell death by apoptosis. Alternative splicing results in multiple transcript variants encoding different isoforms.Spécification
NUP93 | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q66HC5, Q8BJ71, Q8N1F7 | |
93 kDa nucleoporin, KIAA0095, nucleoporin 93kDa, Nucleoporin Nup93, NUP93 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
291874, 71805, 9688 |
Western Blot | |
Unconjugated | |
NUP93 | |
Liquid | |
Nup93 | |
NUP93 Fusion Protein Ag3735 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |