Learn More
p115/USO1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 13509-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16801764
  • 140.00€ / 20µL
Expédition estimée: 15-04-2024
Description
The protein encoded by this gene is a peripheral membrane protein which recycles between the cytosol and the Golgi apparatus during interphase. It is regulated by phosphorylation: dephosphorylated protein associates with the Golgi membrane and dissociates from the membrane upon phosphorylation. Ras-associated protein 1 recruits this protein to coat protein complex II (COPII) vesicles during budding from the endoplasmic reticulum, where it interacts with a set of COPII vesicle-associated SNAREs to form a cis-SNARE complex that promotes targeting to the Golgi apparatus. Transport from the ER to the cis/medial Golgi compartments requires the action of this gene product, GM130 and giantin in a sequential manner.
Spécification
Spécification
p115/USO1
Polyclonal
Unconjugated
USO1
P115, Protein USO1 homolog, TAP, USO1, VDP, Vesicle docking protein
Rabbit
Antigen Affinity Chromatography
RUO
56041, 56042, 8615
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin)
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60763, P41542, Q9Z1Z0
Uso1
p115, USO1 Fusion Protein Ag4431
20 μL
Primary
Mouse, Rat, Human