missing translation for 'onlineSavingsMsg'
Learn More

P2RX7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 11144-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16838023

  • 325.38€ / 150µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The product of this gene belongs to the family of purinoceptors for ATP. This receptor functions as a ligand-gated ion channel and is responsible for ATP-dependent lysis of macrophages through the formation of membrane pores permeable to large molecules. Activation of this nuclear receptor by ATP in the cytoplasm may be a mechanism by which cellular activity can be coupled to changes in gene expression. Multiple alternatively spliced variants have been identified, most of which fit nonsense-mediated decay criteria.
Spécification

Spécification

P2RX7
Polyclonal
Unconjugated
P2RX7
P2RX7, P2X7
Rabbit
Antigen Affinity Chromatography
RUO
18439, 29665, 5027
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Flow Cytometry
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q64663, Q99572, Q9Z1M0
P2rx7
P2RX7 Fusion Protein Ag1600
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions