missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ p53 Transcription Factor Assay Kit (Colorimetric)
Assay Kit (Colorimetric)
Marque: Novus Biologicals™ KA1351
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
Transcription Factor Assay Kit (Colorimetric) | |
Functional | |
TP53 | |
Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53 |
Nuclear Extracts | |
7157 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. |