Learn More
PADI2 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 29169-1-AP-20UL
Informations supplémentaires : Poids : 0.00020kg
Code produit. 17271426
  • 140.00€ / 20µL
Expédition estimée: 02-05-2024
Description
The PADI2 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human PADI2. This antibody recognizes human, mouse, rat antigen. The PADI2 antibody has been validated for the following applications: WB,ELISA analysis.
Spécification
Spécification
PADI2
Polyclonal
Unconjugated
20 μL
11240
Liquid
Western Blot, ELISA
400 μg/mL
Rabbit
Primary
Human, Mouse, Rat