missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAG1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25029-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
PAG (phosphoprotein associated with GEMs), also known as Cbp (Csk-binding protein), is a ubiquitously expressed 46 kDa transmembrane adaptor protein present in membrane rafts (glycosphingolipid-enriched microdomains), which however migrates on SDS PAGE gels anomalously as an 80 kDa molecule. Following tyrosine phosphorylation by Src family kinases, PAG binds and thereby activates the protein tyrosine kinase Csk, the major negative regulator of the Src family kinases. Signaling via the B-cell receptor in B cells or high affinity IgE receptor (FcepsilonRI) in mast cells leads to PAG increased tyrosine phosphorylation and Csk binding, while T cell receptor signaling causes PAG dephosphorylation, loss of Csk binding and increased activation of the protein tyrosine kinase Lck.Spécification
PAG1 | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3U1F9, Q9NWQ8 | |
CBP, Csk binding protein, PAG, PAG1 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
55824, 94212 |
Western Blot | |
Unconjugated | |
PAG1 | |
Liquid | |
PAG1 | |
PAG1 Fusion Protein Ag13393 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Human |