missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAGE3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | PAGE3 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16803095
|
Proteintech
24598-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 13-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16893085
|
Proteintech
24598-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 13-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
PAGE-3 (prostate-associated gene 3 protein), also known as GAGED1 (putative G antigen family Dmember 1), is a 113 amino acid protein that belongsto the GAGE family. The gene that encodes PAGE-3 consists of approximately 6,432 bases andmapsto human chromosome Xp11.21. Chromosome X consists of about 153million base pairs and nearly 1,000 genes. The combination of a X and Y chromosome lead to normal male development while two copies of X lead to normal female development. There are a number ofconditionsrelated to an unusual number and combination ofsexchromosomes being inherited, including Turner′s syndrome, Klinefelter′ssyndrome and Triple X syndrome. Color blindness, hemophilia, and Duchennemuscular dystrophy arewellknown X chromosome-linked conditionswhich affectmalesmore frequently as males carry a single X chromosome.Spécification
PAGE3 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q5JUK9 | |
139793 | |
PAGE3 Fusion Protein Ag20171 | |
Primary | |
-20°C | |
PAGE3 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GAGEC1, GAGED1, PAGE 3, PAGE3 | |
PAGE3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |