missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 51137-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
PAK proteins are critical effectors that link RhoGTPases to cytoskeleton reorganization and nuclear signaling. PAK proteins, a family of serine/threonine p21-activating kinases, include PAK1, PAK2, PAK3 and PAK4. These proteins serve as targets for the small GTP binding proteins Cdc42 and Rac and have been implicated in a wide range of biological activities. PAK1 regulates cell motility and morphology. Alternativelt spliced transcript variants encoding different isoforms have been found for this gene.Spécification
PAK1 | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13153 | |
Alpha PAK, p21 activated kinase 1, p65 PAK, PAK 1, PAK1, PAKalpha | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
5058 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PAK1 | |
Liquid | |
PAK1 | |
Peptide | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |