Learn More
Labconco™ Accessoire pour enceinte sans raccordement Paramount™, support télescopique
For use with Paramount™ Ductless Enclosure. Labconco™ Paramount™ Ductless Enclosure Accessory, Telescoping Base Stand with casters may be adjusted to eight height positions prior to installation.
Marque:  Labconco™ 3746710
Code nomenclature Nacres: HA.23
Informations supplémentaires : Poids : 38.64000kg
Code produit. 15119655
  • 871.84€ / 1 pièce
Pour délai, nous consulter 03 88 67 14 14
Ajouter au panier
Description
Description
  • Durable 4.4cm (1.75 in.) tubular, powder-coated steel frame
  • Casters provide mobility for short distances
  • Telescoping legs are adjustable to eight height positions in 2.54cm (1 in.) increments from 69.8 to 87.6cm (27.5 to 34.5 in.) (must be adjusted prior to installation)
  • Includes four 12.7cm (5 in.) diameter, toe-locking, non-marking polyurethane casters with bearings for quiet operation
  • Models 91.4cm (3 ft.) wide and larger have rear crossbar supports
  • Support Paramount Ductless Enclosures and other select Labconco enclosures
  • Work surface is required (sold separately)
  • Models with four fixed leveling feet also avilable
  • 16-gauge steel accessory shelves are available separately to hook on lower rear cross member for storage and supplies
Spécification
Spécification
Base Stand
73.6cm
Color: Glacier white
ADA
4.4cm (1.75 in.) tubular steel
453.5kg (1000 lb.)
60.9cm
Labconco Paramount Ductless Enclosures, Purifier Class I Enclosures, Purifier Filtered PCR Enclosures, Purifier HEPA Filtered Enclosures, Purifier Vertical Clean Benches, Protector XVS Ventilation Stations, XPert Filtered Balance Stations, XPert & RXPert Filtered Balance Systems
69.8 to 87.6cm (Adjustable in 2.54cm increments)
39kg
Vidéos