Learn More
Abnova™ PARD6B Recombinant Protein
Marque:  Abnova™ H00084612-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16125973
  • 508.00€ / 25µg
Expédition estimée: 27-05-2024
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spécification
Spécification
AAH60847
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
66.66
8
Glutathione Sepharose 4 Fast Flow
25 ug
MNRSHRHGAGSGCLGTMEVKSKFGAEFRRFSLERSKPGKFEEFYGLLQHVHKIPNVDVLVGYADIHGDLLPINNDDNYHKAVSTANPLLRIFIQKKEEADYSAFGTDTLIKKKNVLTNVLRPDNHRKKPHIVISMPQDFRPVSSIIDVDILPETHRRVRLYKYGTEKPLGFYIRDGSSVRVTPHGLEKVPGIFISRLVPGGLAQSTGLLAVNDEVLEVNGIEVSGKSLDQVTDMMIANSRNLIITVRPANQRNNVVRNSRTSGSSGQSTDNSLLGYPQQIEPSFEPEDEDSEEDDIIIEDNGVPQQIPKAVPNTESLESLTQIELSFESGQNGFIPSNEVSLAAIASSSNTEFETHAPDQKLLEEDGTIITL
PAR6B
PARD6B
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
84612
PARD6B (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PARD6B
Human
Recombinant
Solution