Learn More
PARP1 Rabbit, anti-Human, Unconjugated, Clone: 3N19, Proteintech
Rabbit Recombinant Monoclonal Antibody
Marque: Proteintech 80174-1-RR-100UL
Informations supplémentaires : Poids : 0.00020kg
Description
The PARP1 antibody from Proteintech is a rabbit recombinant antibody to a recombinant protein of human PARP1. This antibody recognizes human, mouse, rat antigen. The PARP1 antibody has been validated for the following applications: WB,ELISA analysis.
Spécification
PARP1 | |
Monoclonal | |
250 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse, Rat |
Western Blot, ELISA | |
3N19 | |
Unconjugated | |
100 μL | |
142 | |
Liquid |