missing translation for 'onlineSavingsMsg'
Learn More

PDCD2L Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 20781-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16830355

  • 121.38€ / 20µL
Expédition estimée: 27-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Over expression of PDCD2L attenutates TNF-alpha release in Daudi cells. PDCD2L over-expression restrained proliferation of HEK293T cells. DNA/flow cytometry analysis showed that the over-expression of PDCD2L severely delays cell cycle progression at S phase. These all suggest that PDCD2L might play a role in apoptosis.
Spécification

Spécification

PDCD2L
Polyclonal
Unconjugated
PDCD2L
PDCD2L, programmed cell death 2 like
Rabbit
Antigen Affinity Chromatography
RUO
68079, 84306
-20°C
Liquid
Western Blot
0.82 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8C5N5, Q9BRP1
Pdcd2l
PDCD2L Fusion Protein Ag14795
20 μL
Primary
Human, Mouse
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions