missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGFRL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16217-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
PDGFRL bears significant sequence similarity to the ligand binding domain of platelet-derived growth factor receptor beta. Mutations in the PDGFRL gene, or deletion of a chromosomal segment containing this gene, are associated with sporadic hepatocellular carcinomas, colorectal cancers, and non-small cell lung cancers. This suggests PDGFRL may function as a tumor suppressor.Spécification
PDGFRL | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15198, Q5RJP7, Q6PE55 | |
PDGFR like protein, PDGFRL, PDGRL, PRLTS | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
290771, 5157, 68797 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PDGFRL | |
Liquid | |
Pdgfrl | |
PDGFRL Fusion Protein Ag9218 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |