Learn More
PGAM1 Mouse anti-Human, Unconjugated, Clone: 2H2A9, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67470-1-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16830116
  • 399.00€ / 150µL
Expédition estimée: 12-04-2024
Description
The PGAM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PGAM1. This antibody recognizes human, mouse, rat antigen. The PGAM1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
Spécification
PGAM1
Monoclonal
500 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence
2H2A9
Unconjugated
150 μL
5223
IgG1