missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PI4K2A (PI4K2 alpha), GST Tag Recombinant Protein

Human PI4K2A (PI4K2 alpha), GST Tag Recombinant Protein

846.00€ - 7580.00€

Specifications

Numéro d’adhésion NP_060895
À utiliser avec (application) Kinase Assay
Poids moléculaire 80.3 kDa
Famille des kinases PI3 Kinase
Longueur des protéines Full Length
View More Specs

Products 3
Product Code Brand Quantité Price Quantity & Availability  
Product Code Brand Quantité Price Quantity & Availability  
17168755
missing translation for 'viewDocuments' Promotion Details
Invitrogen™
A32905
100 μg
2435.00€
100µg
View alternative products
missing translation for 'toSaveUpto' 
»
Expédition estimée: 29-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
15950313
missing translation for 'viewDocuments' Promotion Details
Invitrogen™
A31079
20 μg
846.00€
20µg
View alternative products
missing translation for 'toSaveUpto' 
»
Expédition estimée: 15-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
17138805
missing translation for 'viewDocuments' Promotion Details
Invitrogen™
A33390
1 mg
7580.00€
1mg
View alternative products
missing translation for 'toSaveUpto' 
»
Expédition estimée: 29-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

PI4K2A (PI4K2 alpha) lipid kinase or phosphatidylinositol 4-kinase type 2 alpha is involved in the formation of phosphatidylinositolpolyphosphates (PtdInsPs) which are centrally involved in many biologic processes ranging from cell growth and organization of the actin cytoskeleton to endo- and exocytosis. PI4K2A phosphorylates PtdIns at the D-4 position which is an essential step in the biosynthesis of PtdInsPs. PI4K2A is a novel regulator of tumor growth by its action on angiogenesis and HIF-1alpha regulation. PI4K2A generates PtdIns4P-rich domains within the Golgi which regulate targeting of clathrin adaptor AP-1 complexes. PI4K2A is also required for Wnt to regulate LRP6 phosphorylation.
  • Validated for use with the Adapta™ assay
  • Available in multiple pack sizes for convenient use
Specifications

Specifications

NP_060895
80.3 kDa
Full Length
Recombinant
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.
Full Length, Recombinant, Active
Kinase Assay
PI3 Kinase
Human
GST Tag
Dry Ice
Liquid
Videos
SDS
Documents

Documents

Certificates