missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC iota Rabbit anti-Chicken, Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | PKC iota |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16812294
|
Proteintech
13883-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 06-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16802294
|
Proteintech
13883-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 06-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
This gene encodes a member of the protein kinase C (PKC) family of serine/threonine protein kinases. The PKC family comprises at least eight members, which are differentially expressed and are involved in a wide variety of cellular processes. This protein kinase is calcium-independent and phospholipid-dependent. It is not activated by phorbolesters or diacylglycerol. This kinase can be recruited to vesicle tubular clusters (VTCs) by direct interaction with the small GTPase RAB2, where this kinase phosphorylates glyceraldehyde-3-phosphate dehydrogenase (GAPD/GAPDH) and plays a role in microtubule dynamics in the early secretory pathway. This kinase is found to be necessary for BCL-ABL-mediated resistance to drug-induced apoptosis and therefore protects leukemia cells against drug-induced apoptosis. There is a single exon pseudogene mapped on chromosome X.Spécification
PKC iota | |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Chicken, Human | |
P41743 | |
424991, 5584 | |
PKC iota Fusion Protein Ag4990 | |
Primary | |
-20°C | |
PRKCI |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
aPKC lambda/iota, DXS1179E, PKC iota, PKCI, PRKC lambda/iota, PRKCI, Protein kinase C iota type, protein kinase C, iota | |
PRKCI | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |