missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasmolipin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | Plasmolipin |
---|---|
Concentration | 0.2 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16843744
|
Proteintech
14868-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16833744
|
Proteintech
14868-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Plasmolipin is an 18 kDa membrane-bound proteolipid that belongs to the tetraspan (4TM) family, a rapidly growing group of myelin-associated proteins. Many of these proteins could be linked to human hereditary demyelinating neuropathies. Studies indicate that plasmolipin is a component of the cytoskeletal structure and has been localized to the apical surface of tubular epithelial cells. It has been shown that, upon addition to lipid bilayers, plasmolipin induces formation of ion channels that are both voltage-dependent and potassium-specific. Plasmolipin was originally isolated from kidney tissue. Subsequently, it has been shown to be widely expressed, not only being present in kidney, but also in the nervous system, thymus, testis, lung, and thyroid gland.Spécification
Plasmolipin | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P47987, Q9DCU2, Q9Y342 | |
51090, 64364, 67801 | |
Plasmolipin Fusion Protein Ag6425 | |
Primary | |
-20°C | |
PLLP |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Plasma membrane proteolipid, Plasmolipin, PLLP, PMLP, TM4SF11 | |
PLLP | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |