missing translation for 'onlineSavingsMsg'
Learn More

Platine Plasma Étl. Solution, Thermo Scientific Chemicals, Specpure™

Catalog Number p-4408579
Click to view available options
Quantité:
50 mL
100 mL
500 mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
7440-6-6
Pt in 20% HCl
MFCD00011179
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
7440-6-6
Pt in 20% HCl
MFCD00011179

Platinum, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau Platinum
Quantité 50 mL
Odeur Characteristic
Solubilité Miscible with water
Forme physique Liquide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'