missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 55216-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody is specific to PLS3.
Plastins are a family of actin-binding proteins that are conserved throughout eukaryote evolution and expressed in most tissues of higher eukaryotes. In humans, two ubiquitous plastin isoforms (L and T) have been identified. Plastin 1 (otherwise known as Fimbrin) is a third distinct plastin isoform which is specifically expressed at high levels in the small intestine. The L isoform is expressed only in hemopoietic cell lineages, while the T isoform has been found in all other normal cells of solid tissues that have replicative potential (fibroblasts, endothelial cells, epithelial cells, melanocytes, etc.). The C-terminal 570 amino acids of the T-plastin and L-plastin proteins are 83% identical. It contains a potential calcium-binding site near the N terminus. Alternate splicing results in multiple transcript variants.Spécification
PLS3 | |
Polyclonal | |
Unconjugated | |
PLS3 | |
Plastin 3, plastin 3 (T isoform), PLS3, T plastin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
102866, 5358, 81748 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P13797, Q63598, Q99K51 | |
PLS3 | |
Peptide | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |