missing translation for 'onlineSavingsMsg'
Learn More

PMF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 11190-2-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16838103

  • 399.00€ / 150µL
Expédition estimée: 12-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

PMF1 is part of the MIS12 complex which is required for normal chromosome alignment and segregation and kinetochore formation during mitosis. It may act as a cotranscription partner of NFE2L2 involved in regulation of polyamine-induced transcription of SSAT.
Spécification

Spécification

PMF1
Polyclonal
Unconjugated
PMF1
PMF 1, PMF1, polyamine modulated factor 1
Rabbit
Antigen Affinity Chromatography
RUO
11243, 67037
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6P1K2, Q9CPV5
Pmf1
PMF1 Fusion Protein Ag1672
150 μL
Primary
Human, Mouse
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions