missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNCK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13712-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
PNCK is a member of the calcium/calmodulin-dependent protein kinase family of protein serine/threonine kinases (Gardner et al., 2000 ).Spécification
PNCK | |
Polyclonal | |
Unconjugated | |
PNCK | |
BSTK3, CaM KI beta, CaM kinase I beta, CaM kinase IB, CaMK1b, CaMKI beta, PNCK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
139728, 29660, 93843 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O70150, Q6P2M8, Q9QYK9 | |
Pnck | |
PNCK Fusion Protein Ag4650 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |