missing translation for 'onlineSavingsMsg'
Learn More

Chlorhydrate PNU 22394, Tocris

5-HT2C agonist and 5-HT2A/2B partial agonist

Marque:  SPEX Certiprep 2201-10

Code nomenclature Nacres: NB.15

Informations supplémentaires : Poids : 1.00000kg

Quantité. 10mg

 Afficher plus de versions de ce produit

Code produit. 13541190

  • 498.96€ / 10 pièces


Pour délai, nous consulter 03 88 67 14 14
Ajouter au panier

Description

Description

Potent 5-HT2C agonist and partial 5-HT2A/5-HT2B agonist. Non-selective between 5-HT2 receptor subtypes (Ki values are 18, 18 and 66 nM for human recombinant 5-HT2C, 5-HT2A and 5-HT2B receptors respectively). Reduces food intake in rats following subcutaneous administration and displays anorexigenic effects in humans.
Spécification

Identifiants chimiques

15923-42-9
236.743
pnu 22394 hydrochloride, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indole hydrochloride, 6-methyl-1h,2h,3h,4h,5h-azepino 4,5-b indole hydrochloride, 6-methyl-1,2,3,4,5,6-hexahydroazepino 4,5-b indol-6-ium chloride, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indol, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indole-hydrochloride
6-methyl-2,3,4,5-tetrahydro-1H-azepino[4,5-b]indole;hydrochloride
C13H17ClN2
SAIGGEUWSYESTR-UHFFFAOYSA-N
16759162
CN1C2=C(CCNCC2)C3=CC=CC=C31.Cl

Spécification

PNU 22394 hydrochloride
15923-42-9
pnu 22394 hydrochloride, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indole hydrochloride, 6-methyl-1h,2h,3h,4h,5h-azepino 4,5-b indole hydrochloride, 6-methyl-1,2,3,4,5,6-hexahydroazepino 4,5-b indol-6-ium chloride, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indol, 1,2,3,4,5,6-hexahydro-6-methyl-azepino 4,5-b indole-hydrochloride
CN1C2=C(CCNCC2)C3=CC=CC=C31.Cl
236.743
236.74
10mg
C13H17ClN2
SAIGGEUWSYESTR-UHFFFAOYSA-N
6-methyl-2,3,4,5-tetrahydro-1H-azepino[4,5-b]indole;hydrochloride
16759162
>99%
Vidéos