missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2F Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15334-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
POLR2F belongs to the archaeal rpoK/eukaryotic RPB6 RNA polymerase subunit family and is the sixth largest subunit of RNA polymerase II. This polymerase, along with two other DNA-directed RNA polymerases, is responsible for synthesizing messenger RNA in eukaryotes. Three distinct zinc-containing RNA polymerases are found in eukaryotic nuclei: polymerase I for the ribosomal RNA precursor, polymerase II for the mRNA precursor, and polymerase III for 5S and tRNA genes. Each class of RNA polymerase is assembled from 9 to 15 different polypeptides. In yeast, this polymerase subunit, in combination with at least two other subunits, forms a structure that stabilizes the transcribing polymerase on the DNA template.Spécification
POLR2F | |
Polyclonal | |
Unconjugated | |
POLR2F | |
HRBP14.4, POLR2F, POLRF, RPABC14.4, RPABC2, RPB14.4, RPB6, RPB6 homolog, RPC15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5435, 69833, 83503 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88828, P61218, P61219 | |
POLR2F | |
POLR2F Fusion Protein Ag7449 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |