missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR3F Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16044-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The human POLR3F is a component of RNA III polymerase. RNA polymerase III transcribes many essential, small, noncoding RNAs, including the 5S rRNAs and tRNAs. While most pol III-transcribed genes are found scattered throughout the linear chromosome maps or in multiple linear clusters, there is increasing evidence that many of these genes prefer to be spatially clustered, often at or near the nucleolus. This association could create an environment that fosters the coregulation of transcription by pol III with transcription of the large ribosomal RNA repeats by RNA polymerase I (pol I) within the nucleolus. Given the high number of pol III-transcribed genes in all eukaryotic genomes, the spatial organization of these genes is likely to affect a large portion of the other genes in a genome. POLR3F has also been recently identified as an HIV dependency factor (HDF), suggesting that POLR3F may be an important drug target in HIV treatment. At least two isoforms of POLR3F are known to exist.Spécification
POLR3F | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q921X6, Q9H1D9 | |
POLR3F, RNA polymerase III subunit C6, RPC39, RPC6 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
10621, 311487, 70408 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
POLR3F | |
Liquid | |
Polr3f | |
POLR3F Fusion Protein Ag8935 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |