Learn More
Multiroir Controlec™ POLYBOX Storage Drawer Carts
Available in a wide format perfect for A4 type documents and for organizing larger volumes. Multiroir Controlec™ POLYBOX Storage Drawer Carts are made entirely in polypropylene and consequently offer a high resistance to shocks.
Marque: Multiroir Controlec™ 18104
Informations supplémentaires : Poids : 5.00000kg
Description
- Choice of black or white
- Available in a variety of sizes/number of drawers
- Multi-directional casters
- Highly resistant polypropylene material
- Excellent quality/price ratio
Spécification
38 x 30 x 67 cm | |
38 cm | |
67 cm | |
4 | |
4.5 kg |
White | |
Storage of A4 documents and larger volumes | |
Polypropylene | |
6 | |
30 cm |