missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Power Cords for myECL Imager and Pierce G2 Fast Blotter

These electrical power cords have a C/13 connector at one end and a specific plug configuration at the opposite end to match electrical outlets of specific countries.

Marque:  Thermo Scientific™ 84858

Code nomenclature Nacres: NA.32

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 13470987

  • 52.25€ / 1 pièce


Date dexpédition estimée
du fournisseur 08-07-2019
Ajouter au panier

Description

Description

The black, 2.4- to 3.5m-long, cords have been validated to replace the cordset that is supplied with particular Thermo Scientific powered devices. Specifically, these instruments include the myECL Imager (Part No. 62236) and the Pierce G2 Fast Blotter (Part No. 62288).

Highlights:

  • Black
  • 2.4 to 3.5m long
  • C13 connector
  • Country-specific connectors
Spécification

Spécification

Power Cord with C/13 Connector, North America
1 cord
Power Cord with C/13 Connector, North America
Black 3m power cord
Replacement of myECL Imager and Pierce G2 Fast Blotter accessory
Vidéos