missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAR gamma Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16643-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
PPP1R10 is a protein with similarity to a rat protein that has an inhibitory effect on protein phosphatase-1 (PP1). The rat protein localizes to the nucleus and colocalizes with chromatin at distinct phases during mitosis. This gene encodes a protein with similarity to a rat protein that has an inhibitory effect on protein phosphatase-1 (PP1). The rat protein localizes to the nucleus and colocalizes with chromatin at distinct phases during mitosis. This gene lies within the major histocompatibility complex class I region on chromosome 6.Spécification
PPAR gamma | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88275, P37231, P37238 | |
CIMT1, NR1C3, PPAR gamma, PPARG, PPARG1, PPARG2, PPARgamma | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
19016, 25664, 5468 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
PPARG | |
Liquid | |
Pparg | |
PPAR gamma Fusion Protein Ag10005 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |