missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R3B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14190-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Acts as a glycogen-targeting subunit for phosphatase PP1. Facilitates interaction of the PP1 with enzymes of the glycogen metabolism and regulates its activity. Suppresses the rate at which PP1 dephosphorylates (inactivates) glycogen phosphorylase and enhances the rate at which it activates glycogen synthase and therefore limits glycogen breakdown. Its activity is inhibited by PYGL, resulting in inhibition of the glycogen synthase and glycogen phosphorylase phosphatase activities of PP1. Dramatically increases basal and insulin-stimulated glycogen synthesis upon overexpression in hepatocytes.Spécification
PPP1R3B | |
Polyclonal | |
Unconjugated | |
PPP1R3B | |
GL, PP1 subunit R4, PPP1R3B, PPP1R4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
244416, 79660 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86XI6, Q8C767 | |
PPP1R3B | |
PPP1R3B Fusion Protein Ag5521 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |