missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R9B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 55129-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The antibody is specific to PPP1R9B.
interacts with protein phosphatase 1 and actin filaments; involved in regulation of synaptic transmission.Spécification
PPP1R9B | |
0.29 mg/mL | |
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35274, Q6R891, Q96SB3 | |
Neurabin 2, Neurabin II, PPP1R6, PPP1R9, PPP1R9B, SPINO, Spinophilin, Spn | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
217124, 84686, 84687 |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
PPP1R9B | |
Liquid | |
Ppp1r9b | |
Peptide | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |