missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PR Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25871-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The progesterone receptor is a member of the Steroid Hormone Receptor Superfamily. It exists in two distinct isoforms in human cells. PR-B is the transcriptionally active form and is responsible for activating genes for the maintainence of the endometrium, maintece of pregcy, and inhibition of ovulation. PR-A is identical to PR-B except for a 165 amino acid deletion at the N-terminus. This deletion exposes a 140 amino acid inhibitory domain (ID) that acts as a repressor of steroid hormone transcriptional activity. Phospholabeling studies have shown that PR is phosphorylated in at least 9 different locations in response to progestins, which suggests that PR affects many different cellular pathways.Spécification
PR | |
Polyclonal | |
Unconjugated | |
PGR | |
NR3C3, PGR, PR, progesterone receptor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18667, 5241 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P06401, Q00175 | |
Pgr | |
PR Fusion Protein Ag22986 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |